The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016604.3 | NP_057688.2 | 7 - 8 | 1580 | 91 |
XM_011543488.2 | XP_011541790.1 | 7 - 8 | 1295 | 91 | |
XM_011543489.2 | XP_011541791.1 | 7 - 8 | 1754 | 91 | |
XM_017009584.1 | XP_016865073.1 | 3 - 4 | 700 | 91 | |
GenBank mRNA | AB029005.2 | - | 7 - 8 | 1460 | 91 |
AF251039.1 | - | 7 - 8 | 1092 | 91 | |
AF338242.1 | - | 7 - 8 | 1580 | 91 | |
BC000539.2 | - | 4 - 5 | 833 | 91 | |
BC117693.1 | - | 3 - 4 | 752 | 91 | |
BC146788.1 | - | 7 - 8 | 1461 | 91 |