Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC71
Entrez Gene ID
64925
Gene Name
coiled-coil domain containing 71
Gene Aliases
-
Chromosome Location
Chr.3: 49162535 - 49166352 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022903.3 NP_075054.3 1 - 2 87 65
GenBank mRNA AK022862.1 - 1 - 2 58 65
AK098658.1 - 1 - 2 82 65
BC035516.1 - 1 - 2 56 65
DA613715.1 - 1 - 2 92 65

View Full Product Details