Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HEATR5A
Entrez Gene ID
25938
Gene Name
HEAT repeat containing 5A
Gene Aliases
C14orf125
Chromosome Location
Chr.14: 31291788 - 31420582 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015473.3 NP_056288.2 9 - 10 1531 90
GenBank mRNA AK092400.1 - 2 - 3 273 90
BC045764.1 - 3 - 4 544 90
CR749444.1 - 2 - 3 229 90

View Full Product Details