The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015473.3 | NP_056288.2 | 14 - 15 | 2258 | 109 |
GenBank mRNA | AB037737.1 | - | 5 - 6 | 706 | 109 |
AK092400.1 | - | 7 - 8 | 1000 | 109 | |
BC045764.1 | - | 8 - 9 | 1271 | 109 | |
CR749444.1 | - | 7 - 8 | 956 | 109 |