The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015104.2 | NP_055919.2 | 36 - 37 | 5184 | 62 |
XM_005273849.3 | XP_005273906.1 | 36 - 37 | 5157 | 62 | |
XM_005273850.3 | XP_005273907.1 | 36 - 37 | 5151 | 62 | |
XM_011544863.2 | XP_011543165.1 | 36 - 37 | 5181 | 62 | |
XM_011544864.2 | XP_011543166.1 | 36 - 37 | 5166 | 62 | |
XM_011544865.2 | XP_011543167.1 | 36 - 37 | 5174 | 62 | |
XM_011544866.2 | XP_011543168.1 | 36 - 37 | 5181 | 62 | |
XM_011544868.2 | XP_011543170.1 | 21 - 22 | 2795 | 62 | |
GenBank mRNA | AB007864.1 | - | 36 - 37 | 5125 | 62 |
BC008593.1 | - | 7 - 8 | 812 | 62 | |
BC053596.1 | - | 11 - 12 | 1512 | 62 | |
BC110650.1 | - | 36 - 37 | 5184 | 62 |