Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C10orf62
Entrez Gene ID
414157
Gene Name
chromosome 10 open reading frame 62
Gene Aliases
bA548K23.1
Chromosome Location
Chr.10: 97589693 - 97590934 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009997.2 NP_001009997.2 1 - 1 1204 67
GenBank mRNA BC037252.1 - 1 - 1 1170 67
BC043281.1 - 1 - 1 1655 67

View Full Product Details