The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128608.1 | NP_001122080.1 | 17 - 18 | 2203 | 72 |
NM_001265611.1 | NP_001252540.1 | 16 - 17 | 2185 | 72 | |
NM_014994.2 | NP_055809.2 | 16 - 17 | 2185 | 72 | |
NR_049761.1 | 15 - 16 | 1995 | 72 | ||
NR_049762.1 | 14 - 15 | 1946 | 72 | ||
XM_006720438.1 | XP_006720501.1 | 15 - 16 | 2002 | 72 | |
XM_006720439.2 | XP_006720502.1 | 3 - 4 | 277 | 72 | |
XM_011521383.1 | XP_011519685.1 | 16 - 17 | 2020 | 72 | |
XM_011521384.2 | XP_011519686.1 | 17 - 18 | 2167 | 72 | |
XM_017022017.1 | XP_016877506.1 | 15 - 16 | 2296 | 72 | |
GenBank mRNA | AB011168.2 | - | 17 - 18 | 2113 | 72 |
AB177850.1 | - | 8 - 9 | 1033 | 72 | |
AK302482.1 | - | 14 - 15 | 1887 | 72 | |
AL833267.1 | - | 16 - 17 | 2105 | 72 | |
BC036660.1 | - | 15 - 16 | 2000 | 72 | |
BC113983.1 | - | 16 - 17 | 2077 | 72 | |
BC114493.1 | - | 17 - 18 | 2095 | 72 | |
BX640787.1 | - | 13 - 14 | 2350 | 72 | |
BX648786.1 | - | 16 - 17 | 2136 | 72 |