The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015473.3 | NP_056288.2 | 5 - 6 | 783 | 77 |
GenBank mRNA | DC343794.1 | - | 3 - 4 | 315 | 77 |