The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002030.1 | NP_001002030.1 | 5 - 6 | 770 | 85 |
NM_001105544.1 | NP_001099014.1 | 4 - 5 | 548 | 85 | |
NM_001105545.1 | NP_001099015.1 | 3 - 4 | 495 | 85 | |
NM_001139510.1 | NP_001132982.1 | 5 - 6 | 548 | 85 | |
NM_018479.3 | NP_060949.2 | 4 - 5 | 717 | 85 | |
XM_005267047.2 | XP_005267104.1 | 6 - 7 | 695 | 85 | |
XM_005267048.1 | XP_005267105.1 | 5 - 6 | 869 | 85 | |
XM_005267049.2 | XP_005267106.1 | 4 - 5 | 984 | 85 | |
XM_005267050.2 | XP_005267107.1 | 5 - 6 | 610 | 85 | |
XM_011535942.2 | XP_011534244.1 | 6 - 7 | 666 | 85 | |
XM_017011056.1 | XP_016866545.1 | 6 - 7 | 686 | 85 | |
GenBank mRNA | AF220192.1 | - | 4 - 5 | 721 | 85 |
AK021464.1 | - | 1 - 2 | 1042 | 85 | |
AK025760.1 | - | 5 - 6 | 520 | 85 | |
AK025796.1 | - | 5 - 6 | 548 | 85 | |
AK303812.1 | - | 4 - 5 | 316 | 85 | |
AL834469.1 | - | 5 - 6 | 709 | 85 | |
BC003549.1 | - | 5 - 6 | 732 | 85 | |
BE896631.1 | - | 4 - 5 | 400 | 85 | |
BQ224769.1 | - | 4 - 5 | 663 | 85 | |
DA846978.1 | - | 3 - 4 | 435 | 85 | |
EU668355.1 | - | 3 - 4 | 495 | 85 |