The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038603.2 | NP_001033692.2 | 4 - 5 | 1408 | 73 |
NM_001244734.1 | NP_001231663.1 | 3 - 4 | 1372 | 73 | |
XM_005248445.4 | XP_005248502.1 | 4 - 5 | 1702 | 73 | |
XM_005248446.4 | XP_005248503.1 | 4 - 5 | 1496 | 73 | |
XM_005248447.4 | XP_005248504.1 | 3 - 4 | 1667 | 73 | |
GenBank mRNA | AB219936.1 | - | 3 - 4 | 1334 | 73 |
BC033689.1 | - | 4 - 5 | 1393 | 73 | |
DQ682656.1 | - | 4 - 5 | 1393 | 73 | |
DQ682657.1 | - | 3 - 4 | 1357 | 73 | |
DQ682658.1 | - | 4 - 5 | 1004 | 73 |