Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC78
Entrez Gene ID
124093
Gene Name
coiled-coil domain containing 78
Gene Aliases
C16orf25, CNM4, JFP10, hsCCDC78
Chromosome Location
Chr.16: 722578 - 726880 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031737.2 NP_001026907.2 5 - 6 602 69
XM_006720838.1 XP_006720901.1 4 - 5 807 69
XM_006720843.3 XP_006720906.1 5 - 6 964 69
XM_011522357.1 XP_011520659.1 3 - 4 1020 69
XM_011522360.1 XP_011520662.1 3 - 4 987 69
XM_011522368.1 XP_011520670.1 3 - 4 639 69
XM_017022930.1 XP_016878419.1 5 - 6 702 69
GenBank mRNA AK057178.1 - 5 - 6 669 69
AK298111.1 - 5 - 6 566 69
BC042110.1 - 5 - 6 588 69

View Full Product Details