Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCAS3
Entrez Gene ID
54828
Gene Name
BCAS3, microtubule associated cell migration factor
Gene Aliases
GAOB1, MAAB
Chromosome Location
Chr.17: 60677811 - 61392838 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099432.1 NP_001092902.1 1 - 2 103 91
NM_001320470.1 NP_001307399.1 1 - 2 103 91
NM_017679.3 NP_060149.3 1 - 2 103 91
XM_005257472.1 XP_005257529.1 1 - 2 54 91
XM_011524939.1 XP_011523241.1 1 - 2 54 91
XM_011524940.1 XP_011523242.1 1 - 2 54 91
XM_011524941.1 XP_011523243.1 1 - 2 54 91
XM_011524942.2 XP_011523244.1 1 - 2 54 91
XM_011524943.2 XP_011523245.1 1 - 2 54 91
XM_011524944.1 XP_011523246.1 1 - 2 54 91
XM_011524945.2 XP_011523247.1 1 - 2 54 91
XM_011524946.2 XP_011523248.1 1 - 2 54 91
XM_011524947.1 XP_011523249.1 1 - 2 54 91
XM_011524948.2 XP_011523250.1 1 - 2 54 91
XM_011524949.2 XP_011523251.1 1 - 2 54 91
XM_011524950.1 XP_011523252.1 1 - 2 54 91
XM_011524951.2 XP_011523253.1 1 - 2 54 91
XM_017024783.1 XP_016880272.1 1 - 2 54 91
XM_017024784.1 XP_016880273.1 1 - 2 54 91
XM_017024785.1 XP_016880274.1 1 - 2 54 91
XM_017024786.1 XP_016880275.1 1 - 2 54 91
XM_017024787.1 XP_016880276.1 1 - 2 54 91
XM_017024788.1 XP_016880277.1 1 - 2 54 91
XM_017024789.1 XP_016880278.1 1 - 2 54 91
XM_017024790.1 XP_016880279.1 1 - 2 54 91
XM_017024791.1 XP_016880280.1 1 - 2 54 91
XM_017024792.1 XP_016880281.1 1 - 2 54 91
XM_017024793.1 XP_016880282.1 1 - 2 54 91
XM_017024794.1 XP_016880283.1 1 - 2 54 91
XM_017024795.1 XP_016880284.1 1 - 2 54 91
GenBank mRNA AF361219.1 - 1 - 2 42 91
AK225757.1 - 1 - 2 62 91
AK304791.1 - 1 - 2 89 91
AK314744.1 - 1 - 2 48 91
BC017390.1 - 1 - 2 59 91
DA491709.1 - 1 - 2 40 91

View Full Product Details