Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCAS3
Entrez Gene ID
54828
Gene Name
BCAS3, microtubule associated cell migration factor
Gene Aliases
GAOB1, MAAB
Chromosome Location
Chr.17: 60677811 - 61392838 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099432.1 NP_001092902.1 20 - 21 2181 76
NM_001320470.1 NP_001307399.1 19 - 20 2136 76
NM_017679.3 NP_060149.3 19 - 20 2136 76
XM_005257472.1 XP_005257529.1 20 - 21 2132 76
XM_011524939.1 XP_011523241.1 21 - 22 2222 76
XM_011524940.1 XP_011523242.1 20 - 21 2177 76
XM_011524941.1 XP_011523243.1 19 - 20 2087 76
XM_011524942.2 XP_011523244.1 21 - 22 2222 76
XM_011524943.2 XP_011523245.1 20 - 21 2177 76
XM_011524944.1 XP_011523246.1 21 - 22 2222 76
XM_011524945.2 XP_011523247.1 20 - 21 2177 76
XM_011524946.2 XP_011523248.1 21 - 22 2222 76
XM_011524947.1 XP_011523249.1 21 - 22 2222 76
XM_011524948.2 XP_011523250.1 21 - 22 2222 76
XM_011524949.2 XP_011523251.1 21 - 22 2222 76
XM_011524950.1 XP_011523252.1 21 - 22 2222 76
XM_011524951.2 XP_011523253.1 21 - 22 2222 76
XM_017024783.1 XP_016880272.1 21 - 22 2222 76
XM_017024784.1 XP_016880273.1 21 - 22 2222 76
XM_017024785.1 XP_016880274.1 21 - 22 2222 76
XM_017024786.1 XP_016880275.1 20 - 21 2132 76
XM_017024787.1 XP_016880276.1 21 - 22 2222 76
XM_017024788.1 XP_016880277.1 21 - 22 2222 76
XM_017024789.1 XP_016880278.1 21 - 22 2222 76
XM_017024790.1 XP_016880279.1 20 - 21 2132 76
XM_017024791.1 XP_016880280.1 19 - 20 2087 76
XM_017024792.1 XP_016880281.1 20 - 21 2132 76
XM_017024793.1 XP_016880282.1 19 - 20 2087 76
XM_017024794.1 XP_016880283.1 21 - 22 2222 76
XM_017024795.1 XP_016880284.1 20 - 21 2132 76
XM_017024796.1 XP_016880285.1 12 - 13 1558 76
GenBank mRNA AF361219.1 - 20 - 21 2075 76
AK000135.1 - 5 - 6 614 76
AK022526.1 - 11 - 12 1385 76
AK023054.1 - 11 - 12 1483 76
AK025510.1 - 20 - 21 2097 76
AK225757.1 - 19 - 20 2095 76
AK304791.1 - 12 - 13 1410 76
AK314744.1 - 19 - 20 2081 76
AL831895.1 - 6 - 7 652 76
BC001250.1 - 5 - 6 623 76
BC117275.1 - 20 - 21 2103 76
BC143386.1 - 20 - 21 2103 76
BC143387.1 - 19 - 20 2058 76

View Full Product Details