Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- BCAS3
- Entrez Gene ID
-
54828
- Gene Name
- BCAS3, microtubule associated cell migration factor
- Gene Aliases
-
GAOB1, MAAB
- Chromosome Location
-
Chr.17: 60677811 - 61392838 on Build GRCh38
View Full Product Details