The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256673.1 | NP_001243602.1 | 4 - 5 | 768 | 84 |
NM_001324227.1 | NP_001311156.1 | 5 - 6 | 732 | 84 | |
NM_001324228.1 | NP_001311157.1 | 5 - 6 | 796 | 84 | |
NM_001324229.1 | NP_001311158.1 | 4 - 5 | 1223 | 84 | |
NM_001324230.1 | NP_001311159.1 | 6 - 7 | 1049 | 84 | |
NM_004809.4 | NP_004800.2 | 5 - 6 | 918 | 84 | |
XM_005254792.4 | XP_005254849.1 | 5 - 6 | 1822 | 84 | |
GenBank mRNA | AF111800.1 | - | 4 - 5 | 717 | 84 |
AF156557.1 | - | 5 - 6 | 926 | 84 | |
AK075244.1 | - | 5 - 6 | 814 | 84 | |
AK302051.1 | - | 4 - 5 | 723 | 84 | |
BC034379.1 | - | 5 - 6 | 864 | 84 | |
BC065249.1 | - | 5 - 6 | 864 | 84 | |
CR542089.1 | - | 5 - 6 | 793 | 84 |