Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM69
Entrez Gene ID
140691
Gene Name
tripartite motif containing 69
Gene Aliases
HSD-34, HSD34, RNF36, Trif
Chromosome Location
Chr.15: 44736362 - 44767829 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301144.1 NP_001288073.1 1 - 2 347 72
NM_001301145.1 NP_001288074.1 1 - 2 347 72
NM_001301146.1 NP_001288075.1 1 - 2 347 72
NM_080745.4 NP_542783.2 1 - 2 347 72
GenBank mRNA AF302088.1 - 1 - 2 195 72
AF302089.1 - 1 - 2 195 72
AL360161.1 - 1 - 2 182 72
BC033314.1 - 1 - 2 143 72
BI465168.1 - 1 - 2 179 72

View Full Product Details