The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172623.1 | NP_001166094.1 | 2 - 3 | 643 | 91 |
NM_001172624.1 | NP_001166095.1 | 2 - 3 | 643 | 91 | |
NM_002499.3 | NP_002490.2 | 2 - 3 | 643 | 91 | |
XM_005254408.1 | XP_005254465.1 | 3 - 4 | 893 | 91 | |
XM_011521628.2 | XP_011519930.1 | 2 - 3 | 648 | 91 | |
XM_011521629.2 | XP_011519931.1 | 2 - 3 | 642 | 91 | |
XM_011521630.2 | XP_011519932.1 | 2 - 3 | 642 | 91 | |
XM_011521632.2 | XP_011519934.1 | 2 - 3 | 642 | 91 | |
XM_011521633.2 | XP_011519935.1 | 2 - 3 | 642 | 91 | |
XM_017022231.1 | XP_016877720.1 | 2 - 3 | 642 | 91 | |
XM_017022232.1 | XP_016877721.1 | 3 - 4 | 893 | 91 | |
XM_017022233.1 | XP_016877722.1 | 2 - 3 | 642 | 91 | |
XM_017022234.1 | XP_016877723.1 | 3 - 4 | 685 | 91 | |
XM_017022235.1 | XP_016877724.1 | 2 - 3 | 642 | 91 | |
XM_017022236.1 | XP_016877725.1 | 2 - 3 | 642 | 91 | |
XM_017022237.1 | XP_016877726.1 | 2 - 3 | 642 | 91 | |
XM_017022238.1 | XP_016877727.1 | 2 - 3 | 642 | 91 | |
GenBank mRNA | BC117161.1 | - | 2 - 3 | 517 | 91 |
BC143270.1 | - | 2 - 3 | 518 | 91 | |
BC143271.1 | - | 2 - 3 | 517 | 91 | |
U61262.1 | - | 2 - 3 | 587 | 91 | |
U72391.1 | - | 2 - 3 | 577 | 91 |