Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NEO1
- Entrez Gene ID
-
4756
- Gene Name
- neogenin 1
- Gene Aliases
-
IGDCC2, NGN, NTN1R2
- Chromosome Location
-
Chr.15: 73051715 - 73305206 on Build GRCh38
View Full Product Details