The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195059.1 | NP_001181988.1 | 4 - 5 | 465 | 61 |
NM_025201.4 | NP_079477.2 | 5 - 6 | 615 | 61 | |
GenBank mRNA | AF318373.1 | - | 2 - 3 | 213 | 61 |
AF494534.1 | - | 5 - 6 | 564 | 61 | |
AK096203.1 | - | 5 - 6 | 583 | 61 | |
BC008744.2 | - | 4 - 5 | 426 | 61 |