The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040694.1 | NP_001035784.1 | 9 - 10 | 1659 | 121 |
NM_020238.2 | NP_064623.2 | 9 - 10 | 1659 | 121 | |
XM_006718533.2 | XP_006718596.1 | 9 - 10 | 1621 | 121 | |
XM_011544995.2 | XP_011543297.1 | 9 - 10 | 1621 | 121 | |
XM_011544996.2 | XP_011543298.1 | 9 - 10 | 1621 | 121 | |
XM_011544997.2 | XP_011543299.1 | 9 - 10 | 1581 | 121 | |
XM_011544998.2 | XP_011543300.1 | 9 - 10 | 1621 | 121 | |
GenBank mRNA | AF282265.1 | - | 8 - 9 | 1457 | 121 |
AK056195.1 | - | 9 - 10 | 1634 | 121 | |
AK307633.1 | - | 9 - 10 | 1676 | 121 | |
AY714053.1 | - | 8 - 9 | 1457 | 121 | |
BC032678.1 | - | 9 - 10 | 1583 | 121 | |
BC098576.1 | - | 9 - 10 | 1659 | 121 |