Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HACE1
Entrez Gene ID
57531
Gene Name
HECT domain and ankyrin repeat containing E3 ubiquitin protein ligase 1
Gene Aliases
SPPRS
Chromosome Location
Chr.6: 104682670 - 104859919 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321080.1 NP_001308009.1 11 - 12 1552 149
NM_001321083.1 NP_001308012.1 12 - 13 1499 149
NM_001321084.1 NP_001308013.1 11 - 12 1416 149
NM_020771.3 NP_065822.2 12 - 13 1684 149
NR_104424.1 13 - 14 1778 149
XM_017011113.1 XP_016866602.1 12 - 13 1551 149
XM_017011114.1 XP_016866603.1 11 - 12 1370 149
XM_017011115.1 XP_016866604.1 11 - 12 1526 149
XM_017011116.1 XP_016866605.1 11 - 12 1581 149
XM_017011118.1 XP_016866607.1 9 - 10 1104 149
XM_017011119.1 XP_016866608.1 11 - 12 1387 149
XM_017011120.1 XP_016866609.1 10 - 11 1502 149
XM_017011121.1 XP_016866610.1 10 - 11 1469 149
XM_017011122.1 XP_016866611.1 11 - 12 1221 149
GenBank mRNA AB037741.1 - 1 - 2 2431 149
AK131207.1 - 12 - 13 1684 149
AK291760.1 - 12 - 13 1609 149
AK311633.1 - 1 - 2 233 149
BC034982.2 - 12 - 13 1648 149

View Full Product Details