Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADCY7
Entrez Gene ID
113
Gene Name
adenylate cyclase 7
Gene Aliases
AC7
Chromosome Location
Chr.16: 50246158 - 50318135 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114.4 NP_001105.1 20 - 21 2875 67
XM_011522835.2 XP_011521137.1 21 - 22 2976 67
XM_011522836.2 XP_011521138.1 21 - 22 2992 67
XM_011522837.2 XP_011521139.1 21 - 22 2921 67
XM_011522838.2 XP_011521140.1 21 - 22 3125 67
XM_011522839.2 XP_011521141.1 21 - 22 2952 67
XM_011522840.2 XP_011521142.1 21 - 22 2959 67
XM_011522842.2 XP_011521144.1 21 - 22 3022 67
XM_017022895.1 XP_016878384.1 21 - 22 2992 67
XM_017022896.1 XP_016878385.1 18 - 19 2138 67
XM_017022897.1 XP_016878386.1 18 - 19 2157 67
GenBank mRNA BC126271.1 - 20 - 21 2687 67
D25538.1 - 20 - 21 2872 67

View Full Product Details