Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNS4
Entrez Gene ID
84951
Gene Name
tensin 4
Gene Aliases
CTEN, PP14434
Chromosome Location
Chr.17: 40475828 - 40501681 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032865.5 NP_116254.4 10 - 11 2069 57
XM_005257744.1 XP_005257801.1 10 - 11 2145 57
XM_017025236.1 XP_016880725.1 9 - 10 2070 57
GenBank mRNA AF370421.1 - 10 - 11 2065 57
AF417488.1 - 10 - 11 2031 57
AK027856.1 - 8 - 9 1419 57
BC018706.1 - 9 - 10 1515 57

View Full Product Details