Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GSDMA
Entrez Gene ID
284110
Gene Name
gasdermin A
Gene Aliases
FKSG9, GSDM, GSDM1
Chromosome Location
Chr.17: 39962973 - 39977880 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178171.4 NP_835465.2 2 - 3 331 149
XM_006721832.2 XP_006721895.1 2 - 3 284 149
XM_017024502.1 XP_016879991.1 2 - 3 331 149
XM_017024503.1 XP_016879992.1 2 - 3 331 149
XM_017024504.1 XP_016879993.1 2 - 3 331 149
GenBank mRNA AB093591.1 - 1 - 2 224 149
AF307953.1 - 1 - 2 243 149
AK096439.1 - 2 - 3 333 149
BC109197.2 - 2 - 3 293 149

View Full Product Details