The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032430.1 | NP_115806.1 | 9 - 10 | 1135 | 87 |
XM_005259327.3 | XP_005259384.1 | 9 - 10 | 1204 | 87 | |
XM_011527395.2 | XP_011525697.2 | 7 - 8 | 569 | 87 | |
GenBank mRNA | AB058714.1 | - | 8 - 9 | 670 | 87 |
AF479826.1 | - | 11 - 12 | 1033 | 87 | |
AF479827.1 | - | 9 - 10 | 1135 | 87 | |
AL834275.1 | - | 9 - 10 | 758 | 87 | |
AY458602.1 | - | 9 - 10 | 907 | 87 | |
AY505124.1 | - | 9 - 10 | 858 | 87 | |
HQ830199.1 | - | 9 - 10 | 880 | 87 |