The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032192.3 | NP_115568.2 | 1 - 2 | 549 | 116 |
XM_017025216.1 | XP_016880705.1 | 2 - 3 | 900 | 116 | |
GenBank mRNA | AF435972.1 | - | 1 - 2 | 508 | 116 |
AF464196.1 | - | 1 - 2 | 547 | 116 | |
AK123112.1 | - | 1 - 2 | 522 | 116 | |
AK127542.1 | - | 2 - 3 | 413 | 116 | |
AK129537.1 | - | 2 - 3 | 465 | 116 | |
DA136306.1 | - | 1 - 2 | 550 | 116 |