The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001292038.1 | NP_001278967.1 | 4 - 5 | 602 | 52 |
NM_015274.2 | NP_056089.1 | 4 - 5 | 602 | 52 | |
GenBank mRNA | AB023152.2 | - | 4 - 5 | 561 | 52 |
AK301473.1 | - | 4 - 5 | 587 | 52 | |
AL833071.2 | - | 4 - 5 | 575 | 52 | |
BC033307.1 | - | 4 - 5 | 571 | 52 | |
BC094773.1 | - | 4 - 5 | 602 | 52 |