Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAN2B2
Entrez Gene ID
23324
Gene Name
mannosidase alpha class 2B member 2
Gene Aliases
-
Chromosome Location
Chr.4: 6575174 - 6622403 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001292038.1 NP_001278967.1 3 - 4 434 70
NM_015274.2 NP_056089.1 3 - 4 434 70
GenBank mRNA AB023152.2 - 3 - 4 393 70
AK301473.1 - 3 - 4 419 70
AL832232.1 - 3 - 4 406 70
AL833071.2 - 3 - 4 407 70
BC033307.1 - 3 - 4 403 70
BC094773.1 - 3 - 4 434 70

View Full Product Details