Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FCRL2
Entrez Gene ID
79368
Gene Name
Fc receptor like 2
Gene Aliases
CD307b, FCRH2, IFGP4, IRTA4, SPAP1, SPAP1A, SPAP1B, SPAP1C
Chromosome Location
Chr.1: 157745733 - 157777472 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159488.1 NP_001152960.1 4 - 5 655 67
NM_030764.3 NP_110391.2 4 - 5 655 67
XM_006711535.3 XP_006711598.1 4 - 5 995 67
XM_011509974.2 XP_011508276.1 4 - 5 990 67
XM_011509975.2 XP_011508277.1 4 - 5 992 67
XM_011509976.2 XP_011508278.1 4 - 5 991 67
XM_017002316.1 XP_016857805.1 4 - 5 990 67
XM_017002317.1 XP_016857806.1 4 - 5 991 67
XM_017002318.1 XP_016857807.1 4 - 5 992 67
GenBank mRNA AF459633.1 - 4 - 5 643 67
AK301113.1 - 4 - 5 642 67
AY043465.1 - 4 - 5 642 67
AY358130.1 - 4 - 5 645 67

View Full Product Details