The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032865.5 | NP_116254.4 | 1 - 2 | 62 | 78 |
XM_005257744.1 | XP_005257801.1 | 1 - 2 | 141 | 78 | |
XM_017025236.1 | XP_016880725.1 | 1 - 2 | 141 | 78 | |
XM_017025237.1 | XP_016880726.1 | 1 - 2 | 141 | 78 | |
GenBank mRNA | AF370421.1 | - | 1 - 2 | 58 | 78 |
BM710196.1 | - | 1 - 2 | 62 | 78 |