The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135703.2 | NP_001129175.1 | 2 - 3 | 347 | 73 |
NM_013437.4 | NP_038465.1 | 3 - 4 | 404 | 73 | |
GenBank mRNA | AB208957.1 | - | 1 - 2 | 79 | 73 |
AF166350.1 | - | 3 - 4 | 319 | 73 | |
AK289752.1 | - | 3 - 4 | 308 | 73 | |
AK299374.1 | - | 2 - 3 | 344 | 73 | |
AL698796.1 | - | 3 - 4 | 372 | 73 | |
BC032109.2 | - | 3 - 4 | 280 | 73 | |
DC372924.1 | - | 2 - 3 | 344 | 73 |