The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020936.2 | NP_065987.1 | 9 - 10 | 1027 | 110 |
NR_135104.1 | 9 - 10 | 1027 | 110 | ||
NR_135105.1 | 9 - 10 | 1027 | 110 | ||
XM_011538608.2 | XP_011536910.1 | 9 - 10 | 1069 | 110 | |
XM_017019710.1 | XP_016875199.1 | 9 - 10 | 1066 | 110 | |
XM_017019711.1 | XP_016875200.1 | 8 - 9 | 1121 | 110 | |
XM_017019712.1 | XP_016875201.1 | 9 - 10 | 919 | 110 | |
XM_017019713.1 | XP_016875202.1 | 9 - 10 | 1042 | 110 | |
XM_017019714.1 | XP_016875203.1 | 10 - 11 | 1200 | 110 | |
XM_017019715.1 | XP_016875204.1 | 9 - 10 | 1067 | 110 | |
XM_017019716.1 | XP_016875205.1 | 9 - 10 | 1012 | 110 | |
XM_017019717.1 | XP_016875206.1 | 9 - 10 | 964 | 110 | |
XM_017019718.1 | XP_016875207.1 | 7 - 8 | 1382 | 110 | |
XM_017019719.1 | XP_016875208.1 | 9 - 10 | 919 | 110 | |
XM_017019720.1 | XP_016875209.1 | 7 - 8 | 1997 | 110 | |
XM_017019721.1 | XP_016875210.1 | 9 - 10 | 1013 | 110 | |
GenBank mRNA | AB046815.1 | - | 4 - 5 | 2854 | 110 |
AK074219.1 | - | 9 - 10 | 979 | 110 | |
AK131442.1 | - | 6 - 7 | 1266 | 110 | |
AK301043.1 | - | 9 - 10 | 990 | 110 | |
AK307384.1 | - | 9 - 10 | 1003 | 110 | |
AL833756.1 | - | 7 - 8 | 2064 | 110 | |
BC030020.2 | - | 9 - 10 | 971 | 110 |