Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AIG1
Entrez Gene ID
51390
Gene Name
androgen induced 1
Gene Aliases
AIG-1, dJ95L4.1
Chromosome Location
Chr.6: 143059363 - 143343594 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286587.1 NP_001273516.1 6 - 7 697 101
NM_016108.3 NP_057192.2 5 - 6 754 101
XM_011535885.2 XP_011534187.1 4 - 5 619 101
GenBank mRNA AF151861.1 - 6 - 7 102
AF153605.1 - 5 - 6 697 101
AJ420480.1 - 3 - 4 370 101
AK222475.1 - 5 - 6 716 101
BC025278.1 - 5 - 6 685 101
BX538067.1 - 4 - 5 457 101

View Full Product Details