Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC7
Entrez Gene ID
79741
Gene Name
coiled-coil domain containing 7
Gene Aliases
BIOT2, BioT2-A, BioT2-B, BioT2-C, C10orf68
Chromosome Location
Chr.10: 32446082 - 32882864 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321115.1 NP_001308044.1 43 - 44 4943 112
XM_005252593.2 XP_005252650.1 24 - 25 2730 112
XM_006717505.1 XP_006717568.1 41 - 42 4438 112
XM_006717506.1 XP_006717569.1 40 - 41 4330 112
XM_011519663.1 XP_011517965.1 43 - 44 4651 112
XM_011519666.1 XP_011517968.1 42 - 43 4567 112
XM_011519667.1 XP_011517969.1 42 - 43 4567 112
XM_011519668.1 XP_011517970.1 42 - 43 4561 112
XM_011519669.1 XP_011517971.1 42 - 43 4549 112
XM_011519670.1 XP_011517972.1 42 - 43 4543 112
XM_011519671.1 XP_011517973.1 42 - 43 4543 112
XM_011519672.1 XP_011517974.1 42 - 43 4540 112
XM_011519673.1 XP_011517975.1 42 - 43 4537 112
XM_011519674.1 XP_011517976.1 41 - 42 4459 112
XM_011519675.1 XP_011517977.1 41 - 42 4441 112
XM_011519676.1 XP_011517978.1 41 - 42 4429 112
XM_011519677.1 XP_011517979.1 42 - 43 4429 112
XM_011519678.1 XP_011517980.1 41 - 42 4426 112
XM_011519679.1 XP_011517981.1 40 - 41 4120 112
XM_011519680.2 XP_011517982.1 38 - 39 3786 112
XM_011519683.2 XP_011517985.1 32 - 33 3460 112
XM_011519685.1 XP_011517987.1 35 - 36 3668 112
XM_011519687.1 XP_011517989.1 25 - 26 2575 112
XM_017016638.1 XP_016872127.1 43 - 44 4606 112
XM_017016639.1 XP_016872128.1 42 - 43 4540 112
XM_017016640.1 XP_016872129.1 41 - 42 4459 112
XM_017016641.1 XP_016872130.1 38 - 39 4080 112
XM_017016642.1 XP_016872131.1 38 - 39 4153 112
XM_017016643.1 XP_016872132.1 38 - 39 3770 112
XM_017016645.1 XP_016872134.1 37 - 38 3694 112
XM_017016646.1 XP_016872135.1 34 - 35 3581 112
XM_017016648.1 XP_016872137.1 30 - 31 3302 112
XM_017016649.1 XP_016872138.1 30 - 31 3242 112
XM_017016652.1 XP_016872141.1 27 - 28 2705 112
XM_017016653.1 XP_016872142.1 25 - 26 2615 112
GenBank mRNA AA401575.1 - 2 - 3 294 112
AK023093.1 - 22 - 23 2544 112
AK097668.1 - 16 - 17 1684 112

View Full Product Details