Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC7
Entrez Gene ID
79741
Gene Name
coiled-coil domain containing 7
Gene Aliases
BIOT2, BioT2-A, BioT2-B, BioT2-C, C10orf68
Chromosome Location
Chr.10: 32446082 - 32882864 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321115.1 NP_001308044.1 23 - 24 2833 150
XM_005252593.2 XP_005252650.1 4 - 5 620 150
XM_006717505.1 XP_006717568.1 21 - 22 2328 150
XM_006717506.1 XP_006717569.1 20 - 21 2220 150
XM_011519663.1 XP_011517965.1 23 - 24 2541 150
XM_011519664.1 XP_011517966.1 23 - 24 2541 150
XM_011519666.1 XP_011517968.1 23 - 24 2541 150
XM_011519667.1 XP_011517969.1 23 - 24 2541 150
XM_011519668.1 XP_011517970.1 22 - 23 2451 150
XM_011519669.1 XP_011517971.1 22 - 23 2439 150
XM_011519670.1 XP_011517972.1 22 - 23 2433 150
XM_011519671.1 XP_011517973.1 23 - 24 2541 150
XM_011519673.1 XP_011517975.1 23 - 24 2541 150
XM_011519674.1 XP_011517976.1 23 - 24 2541 150
XM_011519675.1 XP_011517977.1 21 - 22 2331 150
XM_011519676.1 XP_011517978.1 23 - 24 2541 150
XM_011519677.1 XP_011517979.1 22 - 23 2319 150
XM_011519679.1 XP_011517981.1 20 - 21 2010 150
XM_011519680.2 XP_011517982.1 18 - 19 1676 150
XM_011519683.2 XP_011517985.1 12 - 13 1350 150
XM_011519684.2 XP_011517986.1 23 - 24 2541 150
XM_011519685.1 XP_011517987.1 15 - 16 1558 150
XM_011519686.2 XP_011517988.1 23 - 24 2541 150
XM_011519687.1 XP_011517989.1 5 - 6 465 150
XM_017016638.1 XP_016872127.1 23 - 24 2541 150
XM_017016639.1 XP_016872128.1 23 - 24 2541 150
XM_017016640.1 XP_016872129.1 23 - 24 2541 150
XM_017016641.1 XP_016872130.1 18 - 19 1970 150
XM_017016642.1 XP_016872131.1 18 - 19 2043 150
XM_017016643.1 XP_016872132.1 18 - 19 1660 150
XM_017016644.1 XP_016872133.1 23 - 24 2541 150
XM_017016645.1 XP_016872134.1 17 - 18 1584 150
XM_017016646.1 XP_016872135.1 14 - 15 1471 150
XM_017016647.1 XP_016872136.1 23 - 24 2541 150
XM_017016648.1 XP_016872137.1 10 - 11 1192 150
XM_017016649.1 XP_016872138.1 10 - 11 1132 150
XM_017016650.1 XP_016872139.1 23 - 24 2541 150
XM_017016651.1 XP_016872140.1 23 - 24 2541 150
XM_017016652.1 XP_016872141.1 7 - 8 595 150
XM_017016653.1 XP_016872142.1 5 - 6 505 150
GenBank mRNA AK023093.1 - 4 - 5 626 150
BC125090.1 - 7 - 8 795 150
BC125091.1 - 3 - 4 265 150

View Full Product Details