Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB38
Entrez Gene ID
23682
Gene Name
RAB38, member RAS oncogene family
Gene Aliases
NY-MEL-1, rrGTPbp
Chromosome Location
Chr.11: 87809490 - 88175505 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022337.2 NP_071732.1 2 - 3 563 68
GenBank mRNA AF136172.1 - 2 - 3 484 68
AF235022.1 - 2 - 3 527 68
AL546159.3 - 2 - 3 563 68
BC015808.1 - 2 - 3 529 68
BT009842.1 - 2 - 3 480 68

View Full Product Details