Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNHG1
Entrez Gene ID
23642
Gene Name
small nucleolar RNA host gene 1
Gene Aliases
LINC00057, NCRNA00057, U22HG, UHG
Chromosome Location
Chr.11: 62851988 - 62855888 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_003098.1 3 - 4 156 61


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BE539085.1 - 2 - 3 279 Chr. 11 - 62852590 - 62855611 [-] on Build GRCh38 61
GenBankmRNA CN386538.1 - 1 - 2 81 Chr. 11 - 62852434 - 62855208 [-] on Build GRCh38 61
GenBankmRNA BQ719301.1 - 2 - 3 119 Chr. 11 - 62852207 - 62855453 [-] on Build GRCh38 61
GenBankmRNA HY165340.1 - 2 - 3 152 Chr. 11 - 62853376 - 62855467 [-] on Build GRCh38 61
GenBankmRNA AA380402.1 - 2 - 3 122 Chr. 11 - 62854518 - 62855456 [-] on Build GRCh38 61
GenBankmRNA CN386546.1 - 2 - 3 120 Chr. 11 - 62853567 - 62855454 [-] on Build GRCh38 61
GenBankmRNA BM762980.1 - 3 - 4 150 Chr. 11 - 62852491 - 62855882 [-] on Build GRCh38 61
GenBankmRNA BG036368.1 - 397 Chr. - - [-] on Build 61
GenBankmRNA BQ690328.1 - 2 - 3 132 Chr. 11 - 62852211 - 62855466 [-] on Build GRCh38 61
GenBankmRNA DA091339.1 - 3 - 4 153 Chr. 11 - 62854529 - 62855885 [-] on Build GRCh38 61
GenBankmRNA BI820763.1 - 2 - 3 112 Chr. 11 - 62853528 - 62855446 [-] on Build GRCh38 61
GenBankmRNA AW963561.1 - 2 - 3 134 Chr. 11 - 62852467 - 62855467 [-] on Build GRCh38 61
GenBankmRNA AL534564.3 - 2 - 3 478 Chr. 11 - 62854236 - 62855812 [-] on Build GRCh38 61
GenBankmRNA BM760031.1 - 3 - 4 153 Chr. 11 - 62852320 - 62855885 [-] on Build GRCh38 61
GenBankmRNA BE250286.1 - 2 - 3 397 Chr. 11 - 62853551 - 62855731 [-] on Build GRCh38 61
GenBankmRNA BE279521.1 - 1 - 2 228 Chr. 11 - 62853551 - 62855305 [-] on Build GRCh38 61
GenBankmRNA AA084277.1 - 2 - 3 131 Chr. 11 - 62854035 - 62855465 [-] on Build GRCh38 61
GenBankmRNA BM747384.1 - 3 - 4 151 Chr. 11 - 62852532 - 62855883 [-] on Build GRCh38 61
GenBankmRNA BI334001.1 - 2 - 3 123 Chr. 11 - 62852208 - 62855457 [-] on Build GRCh38 61
GenBankmRNA CN386555.1 - 2 - 3 136 Chr. 11 - 62852588 - 62855466 [-] on Build GRCh38 61
GenBankmRNA CN337004.1 - 1 - 2 218 Chr. 11 - 62852607 - 62855345 [-] on Build GRCh38 61
GenBankmRNA BF699462.1 - 3 - 4 155 Chr. 11 - 62852368 - 62855886 [-] on Build GRCh38 61
GenBankmRNA BM548627.1 - 147 Chr. - - [-] on Build 61
GenBankmRNA DA959310.1 - 3 - 4 154 Chr. 11 - 62854199 - 62855885 [-] on Build GRCh38 61
GenBankmRNA BG324911.1 - 2 - 3 418 Chr. 11 - 62853094 - 62855746 [-] on Build GRCh38 61
GenBankmRNA AA373864.1 - 2 - 3 126 Chr. 11 - 62854008 - 62855460 [-] on Build GRCh38 61
GenBankmRNA CN386563.1 - 2 - 3 126 Chr. 11 - 62853804 - 62855460 [-] on Build GRCh38 61
GenBankmRNA CN337012.1 - 2 - 3 113 Chr. 11 - 62852233 - 62855447 [-] on Build GRCh38 61
GenBankmRNA CN386537.1 - 2 - 3 126 Chr. 11 - 62852346 - 62855460 [-] on Build GRCh38 61
GenBankmRNA BI772659.1 - 2 - 3 118 Chr. 11 - 62852204 - 62855451 [-] on Build GRCh38 61
GenBankmRNA BE734043.1 - 1 - 2 212 Chr. 11 - 62853556 - 62855283 [-] on Build GRCh38 61
GenBankmRNA BM746460.1 - 3 - 4 153 Chr. 11 - 62852661 - 62855885 [-] on Build GRCh38 61
GenBankmRNA BM772121.1 - 3 - 4 153 Chr. 11 - 62852500 - 62855885 [-] on Build GRCh38 61
GenBankmRNA AW954429.1 - 2 - 3 121 Chr. 11 - 62852468 - 62855455 [-] on Build GRCh38 61
GenBankmRNA DA079221.1 - 3 - 4 153 Chr. 11 - 62853506 - 62855885 [-] on Build GRCh38 61
GenBankmRNA CN386545.1 - 2 - 3 132 Chr. 11 - 62852621 - 62855460 [-] on Build GRCh38 61
GenBankmRNA BM756504.1 - 3 - 4 151 Chr. 11 - 62853073 - 62855883 [-] on Build GRCh38 61
GenBankmRNA BQ934825.1 - 2 - 3 136 Chr. 11 - 62852148 - 62855467 [-] on Build GRCh38 61
GenBankmRNA CX783853.1 - 2 - 3 403 Chr. 11 - 62852595 - 62855737 [-] on Build GRCh38 61
GenBankmRNA BM772480.1 - 3 - 4 153 Chr. 11 - 62852439 - 62855885 [-] on Build GRCh38 61
GenBankmRNA DA363685.1 - 3 - 4 186 Chr. 11 - 62853371 - 62855885 [-] on Build GRCh38 61
GenBankmRNA AA346840.1 - 2 - 3 134 Chr. 11 - 62854041 - 62855467 [-] on Build GRCh38 61
GenBankmRNA BQ954365.1 - 2 - 3 135 Chr. 11 - 62852813 - 62855467 [-] on Build GRCh38 61
GenBankmRNA CN337005.1 - 1 - 2 229 Chr. 11 - 62852593 - 62855353 [-] on Build GRCh38 61
GenBankmRNA DA454668.1 - 3 - 4 150 Chr. 11 - 62854516 - 62855882 [-] on Build GRCh38 61
GenBankmRNA DA374845.1 - 3 - 4 153 Chr. 11 - 62853525 - 62855885 [-] on Build GRCh38 61
GenBankmRNA CN386562.1 - 2 - 3 133 Chr. 11 - 62853803 - 62855467 [-] on Build GRCh38 61
GenBankmRNA BM751992.1 - 3 - 4 151 Chr. 11 - 62852613 - 62855883 [-] on Build GRCh38 61
GenBankmRNA AV728106.1 - 2 - 3 126 Chr. 11 - 62852319 - 62855460 [-] on Build GRCh38 61
GenBankmRNA BG527213.1 - 3 - 4 154 Chr. 11 - 62852292 - 62855885 [-] on Build GRCh38 61
GenBankmRNA CN337013.1 - 1 - 2 47 Chr. 11 - 62853573 - 62855174 [-] on Build GRCh38 61
GenBankmRNA BI754745.1 - 2 - 3 521 Chr. 11 - 62853090 - 62855858 [-] on Build GRCh38 61
GenBankmRNA BX340858.2 - 2 - 3 124 Chr. 11 - 62852184 - 62855458 [-] on Build GRCh38 61
GenBankmRNA DA055802.1 - 3 - 4 151 Chr. 11 - 62854206 - 62855883 [-] on Build GRCh38 61
GenBankmRNA AL559848.3 - 2 - 3 126 Chr. 11 - 62852053 - 62855460 [-] on Build GRCh38 61
GenBankmRNA CB125560.1 - 3 - 4 153 Chr. 11 - 62852832 - 62855885 [-] on Build GRCh38 61
GenBankmRNA BM848355.1 - 3 - 4 153 Chr. 11 - 62852550 - 62855885 [-] on Build GRCh38 61
GenBankmRNA CN386536.1 - 2 - 3 136 Chr. 11 - 62852433 - 62855467 [-] on Build GRCh38 61
GenBankmRNA DB040296.1 - 2 - 3 549 Chr. 11 - 62854905 - 62855882 [-] on Build GRCh38 61
GenBankmRNA BM842784.1 - 3 - 4 153 Chr. 11 - 62852297 - 62855885 [-] on Build GRCh38 61
GenBankmRNA DA972474.1 - 3 - 4 153 Chr. 11 - 62853371 - 62855885 [-] on Build GRCh38 61
GenBankmRNA CN337019.1 - 2 - 3 116 Chr. 11 - 62853035 - 62855450 [-] on Build GRCh38 61
GenBankmRNA AA218776.1 - 2 - 3 115 Chr. 11 - 62853076 - 62855449 [-] on Build GRCh38 61
GenBankmRNA AA377743.1 - 132 Chr. - - [-] on Build 61
GenBankmRNA BG623014.1 - 3 - 4 154 Chr. 11 - 62852274 - 62855885 [-] on Build GRCh38 61
GenBankmRNA BM740049.1 - 2 - 3 148 Chr. 11 - 62852514 - 62855467 [-] on Build GRCh38 61
GenBankmRNA CN386544.1 - 2 - 3 117 Chr. 11 - 62852426 - 62855451 [-] on Build GRCh38 61
GenBankmRNA BE543531.1 - 380 Chr. - - [-] on Build 61
GenBankmRNA BM772174.1 - 3 - 4 153 Chr. 11 - 62852491 - 62855885 [-] on Build GRCh38 61
GenBankmRNA AV656829.1 - 2 - 3 105 Chr. 11 - 62852817 - 62855439 [-] on Build GRCh38 61
GenBankmRNA HY048030.1 - 3 - 4 215 Chr. 11 - 62852631 - 62855947 [-] on Build GRCh38 61
GenBankmRNA BI260697.1 - 2 - 3 143 Chr. 11 - 62852486 - 62855464 [-] on Build GRCh38 61
GenBankmRNA DB258464.1 - 3 - 4 153 Chr. 11 - 62854193 - 62855885 [-] on Build GRCh38 61
GenBankmRNA BM771178.1 - 3 - 4 151 Chr. 11 - 62852441 - 62855883 [-] on Build GRCh38 61
GenBankmRNA BM753954.1 - 3 - 4 154 Chr. 11 - 62852631 - 62855885 [-] on Build GRCh38 61
GenBankmRNA BX445478.2 - 2 - 3 126 Chr. 11 - 62852120 - 62855460 [-] on Build GRCh38 61
GenBankmRNA BF307997.1 - 1 - 2 110 Chr. 11 - 62853094 - 62855216 [-] on Build GRCh38 61
GenBankmRNA BP428502.1 - 1 - 2 89 Chr. 11 - 62853098 - 62855214 [-] on Build GRCh38 61
GenBankmRNA CN386561.1 - 3 - 4 154 Chr. 11 - 62853547 - 62855881 [-] on Build GRCh38 61
GenBankmRNA HY171921.1 - 3 - 4 152 Chr. 11 - 62853430 - 62855884 [-] on Build GRCh38 61
GenBankmRNA CN337010.1 - 1 - 2 106 Chr. 11 - 62853203 - 62855224 [-] on Build GRCh38 61
GenBankmRNA AW958661.1 - 2 - 3 144 Chr. 11 - 62853809 - 62855478 [-] on Build GRCh38 61
GenBankmRNA AL555120.3 - 2 - 3 132 Chr. 11 - 62852324 - 62855466 [-] on Build GRCh38 61
GenBankmRNA AA305276.1 - 2 - 3 126 Chr. 11 - 62853094 - 62855460 [-] on Build GRCh38 61
GenBankmRNA HY109978.1 - 3 - 4 150 Chr. 11 - 62852554 - 62855882 [-] on Build GRCh38 61
GenBankmRNA BQ637146.1 - 2 - 3 136 Chr. 11 - 62852445 - 62855467 [-] on Build GRCh38 61
GenBankmRNA BG823022.1 - 2 - 3 519 Chr. 11 - 62854028 - 62855857 [-] on Build GRCh38 61
GenBankmRNA CN386568.1 - 2 - 3 143 Chr. 11 - 62852447 - 62855466 [-] on Build GRCh38 61
GenBankmRNA BM845735.1 - 3 - 4 150 Chr. 11 - 62852642 - 62855880 [-] on Build GRCh38 61
GenBankmRNA C05029.1 - 1 131 Chr. 20 - 29810020 - 29810503 [+] on Build GRCh38 61
GenBankmRNA BM753151.1 - 3 - 4 153 Chr. 11 - 62852628 - 62855885 [-] on Build GRCh38 61
GenBankmRNA BU196426.1 - 149 Chr. - - [-] on Build 61
GenBankmRNA BY996166.1 - 2 - 3 144 Chr. 11 - 62852414 - 62855467 [-] on Build GRCh38 61
GenBankmRNA DA380067.1 - 3 - 4 153 Chr. 11 - 62854202 - 62855885 [-] on Build GRCh38 61
GenBankmRNA CN386543.1 - 2 - 3 126 Chr. 11 - 62852444 - 62855460 [-] on Build GRCh38 61
GenBankmRNA BF132715.1 - 134 Chr. - - [-] on Build 61
GenBankmRNA BE278295.1 - 1 - 2 170 Chr. 11 - 62853551 - 62855299 [-] on Build GRCh38 61
GenBankmRNA CK826165.1 - 2 - 3 270 Chr. 11 - 62852643 - 62855599 [-] on Build GRCh38 61
GenBankmRNA DA268444.1 - 3 - 4 150 Chr. 11 - 62854183 - 62855882 [-] on Build GRCh38 61
GenBankmRNA BQ477769.1 - 2 - 3 270 Chr. 11 - 62852646 - 62855599 [-] on Build GRCh38 61
GenBankmRNA CN386552.1 - 2 - 3 126 Chr. 11 - 62852374 - 62855460 [-] on Build GRCh38 61
GenBankmRNA BM766186.1 - 3 - 4 153 Chr. 11 - 62852453 - 62855885 [-] on Build GRCh38 61
GenBankmRNA AA312093.1 - 2 - 3 115 Chr. 11 - 62853911 - 62855449 [-] on Build GRCh38 61
GenBankmRNA BE873032.1 - 382 Chr. - - [-] on Build 61
GenBankmRNA DA564817.1 - 3 - 4 429 Chr. 11 - 62854470 - 62855882 [-] on Build GRCh38 61
GenBankmRNA BI768992.1 - 2 - 3 127 Chr. 11 - 62852281 - 62855460 [-] on Build GRCh38 61
GenBankmRNA BE792700.1 - 2 - 3 110 Chr. 11 - 62853551 - 62855444 [-] on Build GRCh38 61
GenBankmRNA N76275.1 - 1 - 2 94 Chr. 11 - 62852435 - 62855216 [-] on Build GRCh38 61
GenBankmRNA CR992196.1 - 2 - 3 140 Chr. 11 - 62852199 - 62855467 [-] on Build GRCh38 61
GenBankmRNA BM765544.1 - 3 - 4 153 Chr. 11 - 62852615 - 62855885 [-] on Build GRCh38 61
GenBankmRNA HY069205.1 - 3 - 4 150 Chr. 11 - 62853407 - 62855882 [-] on Build GRCh38 61
GenBankmRNA AW958656.1 - 144 Chr. - - [-] on Build 61
GenBankmRNA BM011342.1 - 2 - 3 114 Chr. 11 - 62853094 - 62855446 [-] on Build GRCh38 61
GenBankmRNA BM746222.1 - 3 - 4 153 Chr. 11 - 62852614 - 62855885 [-] on Build GRCh38 61
GenBankmRNA CN386567.1 - 2 - 3 139 Chr. 11 - 62854036 - 62855466 [-] on Build GRCh38 61
GenBankmRNA DA849223.1 - 3 - 4 153 Chr. 11 - 62853354 - 62855885 [-] on Build GRCh38 61
GenBankmRNA CN386542.1 - 2 - 3 136 Chr. 11 - 62852467 - 62855467 [-] on Build GRCh38 61
GenBankmRNA BI253434.1 - 2 - 3 133 Chr. 11 - 62852193 - 62855467 [-] on Build GRCh38 61
GenBankmRNA BX397597.2 - 8 - 9 Chr. 11 - 62852022 - 62855463 [+] on Build GRCh38 61
GenBankmRNA BM710724.1 - 2 - 3 134 Chr. 11 - 62852533 - 62855467 [-] on Build GRCh38 61
GenBankmRNA BM747357.1 - 3 - 4 151 Chr. 11 - 62852529 - 62855883 [-] on Build GRCh38 61
GenBankmRNA BX419062.2 - 2 - 3 134 Chr. 11 - 62852413 - 62855464 [-] on Build GRCh38 61
GenBankmRNA BF308905.1 - 1 - 2 116 Chr. 11 - 62853094 - 62855216 [-] on Build GRCh38 61
GenBankmRNA CN386550.1 - 2 - 3 104 Chr. 11 - 62852577 - 62855438 [-] on Build GRCh38 61
GenBankmRNA BG614836.1 - 3 - 4 152 Chr. 11 - 62852245 - 62855883 [-] on Build GRCh38 61
GenBankmRNA BI768134.1 - 2 - 3 126 Chr. 11 - 62852465 - 62855460 [-] on Build GRCh38 61
GenBankmRNA DA727570.1 - 3 - 4 153 Chr. 11 - 62852469 - 62855885 [-] on Build GRCh38 61
GenBankmRNA BI255462.1 - 2 - 3 135 Chr. 11 - 62852217 - 62855467 [-] on Build GRCh38 61
GenBankmRNA CN386551.1 - 2 - 3 140 Chr. 11 - 62852460 - 62855474 [-] on Build GRCh38 61
GenBankmRNA BE779013.1 - 2 - 3 504 Chr. 11 - 62854016 - 62855847 [-] on Build GRCh38 61
GenBankmRNA BM710632.1 - 1 - 2 72 Chr. 11 - 62853098 - 62855199 [-] on Build GRCh38 61
GenBankmRNA DA873932.1 - 2 - 3 548 Chr. 11 - 62854904 - 62855882 [-] on Build GRCh38 61
GenBankmRNA BX337289.2 - 2 - 3 133 Chr. 11 - 62851994 - 62855467 [-] on Build GRCh38 61
GenBankmRNA BM762312.1 - 3 - 4 153 Chr. 11 - 62853073 - 62855885 [-] on Build GRCh38 61
GenBankmRNA BG826947.1 - 2 - 3 106 Chr. 11 - 62853094 - 62855439 [-] on Build GRCh38 61
GenBankmRNA BM741778.1 - 3 - 4 153 Chr. 11 - 62852450 - 62855885 [-] on Build GRCh38 61
GenBankmRNA BE794987.1 - 1 - 2 106 Chr. 11 - 62853551 - 62855216 [-] on Build GRCh38 61
GenBankmRNA CN386559.1 - 2 - 3 142 Chr. 11 - 62852641 - 62855460 [-] on Build GRCh38 61
GenBankmRNA BI755677.1 - 2 - 3 515 Chr. 11 - 62853073 - 62855849 [-] on Build GRCh38 61
GenBankmRNA BM765988.1 - 3 - 4 153 Chr. 11 - 62852538 - 62855885 [-] on Build GRCh38 61
GenBankmRNA DB272428.1 - 3 - 4 153 Chr. 11 - 62854208 - 62855885 [-] on Build GRCh38 61
GenBankmRNA BI258011.1 - 2 - 3 280 Chr. 11 - 62852622 - 62855615 [-] on Build GRCh38 61
GenBankmRNA BM841851.1 - 3 - 4 153 Chr. 11 - 62852325 - 62855885 [-] on Build GRCh38 61
GenBankmRNA BI835258.1 - 2 - 3 135 Chr. 11 - 62853179 - 62855466 [-] on Build GRCh38 61
GenBankmRNA CN337018.1 - 2 - 3 293 Chr. 11 - 62852291 - 62855621 [-] on Build GRCh38 61
GenBankmRNA DA679485.1 - 3 - 4 150 Chr. 11 - 62854614 - 62855882 [-] on Build GRCh38 61
GenBankmRNA BM853709.1 - 2 - 3 126 Chr. 11 - 62854016 - 62855460 [-] on Build GRCh38 61
GenBankmRNA BF149374.1 - 3 - 4 Chr. 11 - 62854016 - 62855460 [+] on Build GRCh38 61
GenBankmRNA CN386549.1 - 2 - 3 529 Chr. 11 - 62854042 - 62855860 [-] on Build GRCh38 61
GenBankmRNA CB997695.1 - 180 Chr. - - [-] on Build 61
GenBankmRNA R57509.1 - 71 Chr. - - [-] on Build 61
GenBankmRNA BM772875.1 - 3 - 4 153 Chr. 11 - 62852441 - 62855885 [-] on Build GRCh38 61
GenBankmRNA BG828516.1 - 2 - 3 113 Chr. 11 - 62853094 - 62855446 [-] on Build GRCh38 61
GenBankmRNA BF205426.1 - 2 - 3 122 Chr. 11 - 62853094 - 62855451 [-] on Build GRCh38 61
GenBankmRNA DA913848.1 - 3 - 4 153 Chr. 11 - 62854194 - 62855885 [-] on Build GRCh38 61
GenBankmRNA BI085491.1 - 2 - 3 137 Chr. 11 - 62853094 - 62855467 [-] on Build GRCh38 61
GenBankmRNA BM979103.1 - 2 - 3 553 Chr. 11 - 62853575 - 62855887 [-] on Build GRCh38 61
GenBankmRNA CB996795.1 - 3 - 4 180 Chr. 11 - 62853090 - 62855885 [-] on Build GRCh38 61
GenBankmRNA DA382593.1 - 3 - 4 153 Chr. 11 - 62854442 - 62855885 [-] on Build GRCh38 61
GenBankmRNA BM771760.1 - 2 - 3 146 Chr. 11 - 62852449 - 62855467 [-] on Build GRCh38 61
GenBankmRNA BM823449.1 - 2 - 3 124 Chr. 11 - 62852446 - 62855458 [-] on Build GRCh38 61
GenBankmRNA BE733161.1 - 1 - 2 222 Chr. 11 - 62853551 - 62855312 [-] on Build GRCh38 61
GenBankmRNA DA728354.1 - 3 - 4 151 Chr. 11 - 62852592 - 62855883 [-] on Build GRCh38 61
GenBankmRNA BM847417.1 - 3 - 4 153 Chr. 11 - 62852950 - 62855885 [-] on Build GRCh38 61
GenBankmRNA BM770809.1 - 3 - 4 153 Chr. 11 - 62852636 - 62855885 [-] on Build GRCh38 61
GenBankmRNA DA177942.1 - 3 - 4 153 Chr. 11 - 62854158 - 62855885 [-] on Build GRCh38 61
GenBankmRNA AA082320.1 - 2 - 3 124 Chr. 11 - 62852394 - 62855458 [-] on Build GRCh38 61
GenBankmRNA AA310310.1 - 2 - 3 126 Chr. 11 - 62852808 - 62855460 [-] on Build GRCh38 61
GenBankmRNA BG723036.1 - 3 - 4 436 Chr. 11 - 62853560 - 62855885 [-] on Build GRCh38 61
GenBankmRNA BM846098.1 - 3 - 4 150 Chr. 11 - 62852669 - 62855882 [-] on Build GRCh38 61
GenBankmRNA BX091505.1 - 2 - 3 126 Chr. 11 - 62852395 - 62855460 [-] on Build GRCh38 61
GenBankmRNA CF456049.1 - 2 - 3 159 Chr. 11 - 62852298 - 62855460 [-] on Build GRCh38 61
GenBankmRNA CN337014.1 - 288 Chr. - - [-] on Build 61
GenBankmRNA AL514896.3 - 2 - 3 110 Chr. 11 - 62852158 - 62855444 [-] on Build GRCh38 61
GenBankmRNA BG681844.1 - 2 - 3 127 Chr. 11 - 62852264 - 62855451 [-] on Build GRCh38 61
GenBankmRNA DB126555.1 - 3 - 4 150 Chr. 11 - 62854205 - 62855882 [-] on Build GRCh38 61
GenBankmRNA CN386565.1 - 2 - 3 116 Chr. 11 - 62852818 - 62855450 [-] on Build GRCh38 61
GenBankmRNA BU553089.1 - 2 - 3 158 Chr. 11 - 62852439 - 62855467 [-] on Build GRCh38 61
GenBankmRNA CX760106.1 - 2 - 3 532 Chr. 11 - 62853804 - 62855866 [-] on Build GRCh38 61
GenBankmRNA H84684.1 - 2 - 3 139 Chr. 11 - 62853547 - 62855467 [-] on Build GRCh38 61
GenBankmRNA DA609299.1 - 3 - 4 153 Chr. 11 - 62853371 - 62855885 [-] on Build GRCh38 61
GenBankmRNA BM011936.1 - 2 - 3 114 Chr. 11 - 62853094 - 62855446 [-] on Build GRCh38 61
GenBankmRNA AV654083.1 - 2 - 3 135 Chr. 11 - 62852817 - 62855467 [-] on Build GRCh38 61
GenBankmRNA BM765264.1 - 3 - 4 153 Chr. 11 - 62852483 - 62855885 [-] on Build GRCh38 61
GenBankmRNA BE794379.1 - 2 - 3 107 Chr. 11 - 62853551 - 62855441 [-] on Build GRCh38 61
GenBankmRNA CN386539.1 - 2 - 3 127 Chr. 11 - 62852439 - 62855457 [-] on Build GRCh38 61
GenBankmRNA BX341807.2 - 2 - 3 133 Chr. 11 - 62852048 - 62855467 [-] on Build GRCh38 61
GenBankmRNA BM747810.1 - 3 - 4 151 Chr. 11 - 62853561 - 62855883 [-] on Build GRCh38 61
GenBankmRNA BE744456.1 - 110 Chr. - - [-] on Build 61
GenBankmRNA BF205416.1 - 1 - 2 102 Chr. 11 - 62853094 - 62855216 [-] on Build GRCh38 61
GenBankmRNA BI755575.1 - 2 - 3 511 Chr. 11 - 62852822 - 62855849 [-] on Build GRCh38 61
GenBankmRNA CB268843.1 - 1 - 2 38 Chr. 11 - 62852419 - 62855165 [-] on Build GRCh38 61
GenBankmRNA CN386547.1 - 2 - 3 131 Chr. 11 - 62852444 - 62855460 [-] on Build GRCh38 61
GenBankmRNA BI255546.1 - 2 - 3 138 Chr. 11 - 62852267 - 62855462 [-] on Build GRCh38 61
GenBankmRNA CN337007.1 - 2 - 3 126 Chr. 11 - 62852749 - 62855460 [-] on Build GRCh38 61
GenBankmRNA BE545829.1 - 2 - 3 116 Chr. 11 - 62852235 - 62855450 [-] on Build GRCh38 61
GenBankmRNA AA808004.1 - 2 - 3 427 Chr. 11 - 62854905 - 62855758 [-] on Build GRCh38 61
GenBankmRNA BU168780.1 - 2 - 3 149 Chr. 11 - 62852268 - 62855483 [-] on Build GRCh38 61
GenBankmRNA BI911700.1 - 137 Chr. - - [-] on Build 61
GenBankmRNA CN386556.1 - 2 - 3 126 Chr. 11 - 62852481 - 62855460 [-] on Build GRCh38 61
GenBankmRNA L36587.3 - 2 - 3 396 Chr. 11 - 62851989 - 62855730 [-] on Build GRCh38 61
GenBankmRNA DA380033.1 - 3 - 4 150 Chr. 11 - 62852545 - 62855882 [-] on Build GRCh38 61
GenBankmRNA BM841594.1 - 3 - 4 153 Chr. 11 - 62852299 - 62855885 [-] on Build GRCh38 61
GenBankmRNA AA380722.1 - 2 - 3 134 Chr. 11 - 62853556 - 62855467 [-] on Build GRCh38 61
GenBankmRNA AA401593.1 - 2 - 3 111 Chr. 11 - 62852604 - 62855445 [-] on Build GRCh38 61
GenBankmRNA CA313374.1 - 2 - 3 552 Chr. 11 - 62854016 - 62855887 [-] on Build GRCh38 61
GenBankmRNA BF346105.1 - 2 - 3 173 Chr. 11 - 62854080 - 62855467 [-] on Build GRCh38 61
GenBankmRNA BM843108.1 - 3 - 4 153 Chr. 11 - 62852291 - 62855885 [-] on Build GRCh38 61
GenBankmRNA CN386564.1 - 2 - 3 138 Chr. 11 - 62852567 - 62855460 [-] on Build GRCh38 61
GenBankmRNA DA350333.1 - 3 - 4 148 Chr. 11 - 62854174 - 62855880 [-] on Build GRCh38 61
GenBankmRNA BE250260.1 - 2 - 3 397 Chr. 11 - 62852593 - 62855731 [-] on Build GRCh38 61


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream small nucleolar RNA, C/D box 27 SNORD27 RNU27, U27 Sense Chr. - 62855012 - 62855083 on Build GRCh38
Downstream FSHD region gene 1 family member B, pseudogene FRG1BP C20orf80, FRG1B, bA348I14.2 566.7kb Sense Chr. - 30377203 - 30399331 on Build GRCh38

View Full Product Details