The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000459.4 | NP_000450.2 | 13 - 14 | 2659 | 118 |
NM_001290077.1 | NP_001277006.1 | 12 - 13 | 2530 | 118 | |
NM_001290078.1 | NP_001277007.1 | 11 - 12 | 2218 | 118 | |
XM_005251561.1 | XP_005251618.1 | 13 - 14 | 2326 | 118 | |
XM_005251563.1 | XP_005251620.1 | 12 - 13 | 2197 | 118 | |
GenBank mRNA | AB086825.1 | - | 12 - 13 | 2186 | 118 |
AB208796.1 | - | 13 - 14 | 2337 | 118 | |
AK291775.1 | - | 13 - 14 | 2347 | 118 | |
AK294887.1 | - | 11 - 12 | 2219 | 118 | |
AK295043.1 | - | 12 - 13 | 2256 | 118 | |
BC035514.1 | - | 13 - 14 | 2334 | 118 | |
L06139.1 | - | 13 - 14 | 2357 | 118 |