Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TEK
Entrez Gene ID
7010
Gene Name
TEK receptor tyrosine kinase
Gene Aliases
CD202B, TIE-2, TIE2, VMCM, VMCM1
Chromosome Location
Chr.9: 27109141 - 27230178 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000459.4 NP_000450.2 2 - 3 812 83
NM_001290077.1 NP_001277006.1 2 - 3 812 83
XM_005251561.1 XP_005251618.1 2 - 3 479 83
XM_005251563.1 XP_005251620.1 2 - 3 479 83
GenBank mRNA AB086825.1 - 2 - 3 468 83
AB208796.1 - 2 - 3 490 83
AK291775.1 - 2 - 3 500 83
AK295043.1 - 2 - 3 538 83
BC035514.1 - 2 - 3 487 83
EU826591.1 - 2 - 3 362 83
EU826592.1 - 2 - 3 362 83
L06139.1 - 2 - 3 510 83

View Full Product Details