The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014160.4 | NP_054879.3 | 1 - 2 | 164 | 59 |
GenBank mRNA | AF302084.1 | - | 1 - 2 | 78 | 59 |
AK091318.1 | - | 1 - 2 | 109 | 59 | |
BC015715.1 | - | 1 - 2 | 106 | 59 | |
DR157266.1 | - | 1 - 2 | 164 | 59 |