The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130823.2 | NP_001124295.1 | 29 - 30 | 3348 | 70 |
NM_001318730.1 | NP_001305659.1 | 28 - 29 | 3219 | 70 | |
NM_001318731.1 | NP_001305660.1 | 29 - 30 | 3227 | 70 | |
NM_001379.3 | NP_001370.1 | 28 - 29 | 3300 | 70 | |
GenBank mRNA | AB209413.2 | - | 28 - 29 | 3026 | 70 |
AF180682.1 | - | 26 - 27 | 2807 | 70 | |
BC092517.1 | - | 29 - 30 | 3107 | 70 | |
BC126227.1 | - | 29 - 30 | 3292 | 70 | |
BC144093.1 | - | 29 - 30 | 3292 | 70 | |
X63692.1 | - | 28 - 29 | 3301 | 70 |