The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004333.4 | NP_001004333.2 | 1 - 2 | 344 | 101 |
GenBank mRNA | AK289930.1 | - | 1 - 2 | 127 | 101 |
AM746459.1 | - | 1 - 2 | 111 | 101 | |
BC051802.1 | - | 1 - 2 | 115 | 101 | |
BC062705.1 | - | 1 - 2 | 119 | 101 | |
BM835723.1 | - | 1 - 2 | 344 | 101 | |
CB160051.1 | - | 1 - 2 | 282 | 101 |