The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164269.1 | NP_001157741.1 | 7 - 8 | 790 | 71 |
NM_001164270.1 | NP_001157742.1 | 7 - 8 | 942 | 71 | |
NM_002875.4 | NP_002866.2 | 7 - 8 | 942 | 71 | |
NM_133487.3 | NP_597994.3 | 7 - 8 | 945 | 71 | |
XM_006720626.3 | XP_006720689.1 | 7 - 8 | 993 | 71 | |
XM_011521857.2 | XP_011520159.1 | 7 - 8 | 692 | 71 | |
XM_011521858.2 | XP_011520160.1 | 7 - 8 | 739 | 71 | |
XM_011521859.2 | XP_011520161.1 | 7 - 8 | 876 | 71 | |
XM_011521860.2 | XP_011520162.1 | 7 - 8 | 787 | 71 | |
XM_011521861.2 | XP_011520163.1 | 7 - 8 | 692 | 71 | |
XM_011521862.2 | XP_011520164.1 | 6 - 7 | 792 | 71 | |
GenBank mRNA | AK131299.1 | - | 7 - 8 | 790 | 71 |
AK291969.1 | - | 7 - 8 | 900 | 71 | |
AK313503.1 | - | 7 - 8 | 897 | 71 | |
BC001459.2 | - | 5 - 6 | 608 | 71 | |
BT019705.1 | - | 6 - 7 | 643 | 71 | |
CR536559.1 | - | 6 - 7 | 643 | 71 | |
D13804.1 | - | 7 - 8 | 854 | 71 | |
D14134.1 | - | 7 - 8 | 875 | 71 | |
EU362635.1 | - | 7 - 8 | 942 | 71 |