The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000893.3 | NP_000884.1 | 8 - 9 | 1271 | 63 |
NM_001102416.2 | NP_001095886.1 | 8 - 9 | 1271 | 63 | |
NM_001166451.1 | NP_001159923.1 | 7 - 8 | 1163 | 63 | |
GenBank mRNA | AK223589.1 | - | 8 - 9 | 1220 | 63 |
AK290839.1 | - | 8 - 9 | 1170 | 63 | |
AK298440.1 | - | 9 - 10 | 1185 | 63 | |
AK303768.1 | - | 8 - 9 | 1251 | 63 | |
AK315230.1 | - | 8 - 9 | 1223 | 63 | |
BC026253.1 | - | 8 - 9 | 949 | 63 | |
BC060039.1 | - | 8 - 9 | 1174 | 63 | |
CB148843.1 | - | 1 - 2 | 85 | 63 | |
CD014130.1 | - | 7 - 8 | 1125 | 63 | |
K02566.1 | - | 8 - 9 | 1086 | 63 |