Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KNG1
Entrez Gene ID
3827
Gene Name
kininogen 1
Gene Aliases
BDK, BK, KNG
Chromosome Location
Chr.3: 186717309 - 186744410 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000893.3 NP_000884.1 9 - 10 1363 82
NM_001102416.2 NP_001095886.1 9 - 10 1363 82
NM_001166451.1 NP_001159923.1 8 - 9 1255 82
GenBank mRNA AK223589.1 - 9 - 10 1312 82
AK290839.1 - 9 - 10 1262 82
AK298440.1 - 10 - 11 1277 82
AK303768.1 - 9 - 10 1343 82
AK315230.1 - 9 - 10 1315 82
BC026253.1 - 9 - 10 1041 82
BC060039.1 - 9 - 10 1266 82
CB148843.1 - 2 - 3 177 82
CD014130.1 - 8 - 9 1217 82
K02566.1 - 9 - 10 1178 82

View Full Product Details