The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000893.3 | NP_000884.1 | 10 - 11 | 1442 | 102 |
NM_001166451.1 | NP_001159923.1 | 9 - 10 | 1334 | 102 | |
GenBank mRNA | AK223589.1 | - | 10 - 11 | 1391 | 102 |
AK290839.1 | - | 10 - 11 | 1341 | 102 | |
AK298440.1 | - | 11 - 12 | 1356 | 102 | |
AK315230.1 | - | 10 - 11 | 1394 | 102 | |
BC026253.1 | - | 10 - 11 | 1120 | 102 | |
BC060039.1 | - | 10 - 11 | 1345 | 102 | |
CD014130.1 | - | 9 - 10 | 1296 | 102 | |
K02566.1 | - | 10 - 11 | 1257 | 102 |