The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000775.3 | NP_000766.2 | 5 - 6 | 912 | 108 |
NR_134981.1 | 5 - 6 | 912 | 108 | ||
NR_134982.1 | 5 - 6 | 912 | 108 | ||
GenBank mRNA | AB080265.1 | - | 5 - 6 | 865 | 108 |
AF144566.1 | - | 5 - 6 | 767 | 108 | |
AK315387.1 | - | 5 - 6 | 880 | 108 | |
BC032594.1 | - | 5 - 6 | 915 | 108 | |
CD013972.1 | - | 5 - 6 | 875 | 108 | |
CD013973.1 | - | 5 - 6 | 875 | 108 | |
U37143.2 | - | 5 - 6 | 865 | 108 |