Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CXorf56
Entrez Gene ID
63932
Gene Name
chromosome X open reading frame 56
Gene Aliases
-
Chromosome Location
Chr.X: 119538149 - 119565434 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170570.1 NP_001164041.1 1 - 2 204 58
NM_022101.3 NP_071384.1 1 - 2 204 58
GenBank mRNA AK298877.1 - 1 - 2 182 58
BC023506.2 - 1 - 2 165 58
BQ223940.1 - 1 - 2 169 58
DA921725.1 - 1 - 2 179 58
DB033119.1 - 1 - 2 219 58

View Full Product Details