The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024953.3 | NP_079229.2 | 10 - 11 | 1092 | 122 |
XM_005253938.1 | XP_005253995.1 | 9 - 10 | 938 | 122 | |
XM_006719606.2 | XP_006719669.1 | 10 - 11 | 1752 | 122 | |
XM_017019977.1 | XP_016875466.1 | 8 - 9 | 808 | 122 | |
GenBank mRNA | AB054990.1 | - | 10 - 11 | 1034 | 122 |
AK023151.1 | - | 7 - 8 | 737 | 122 | |
AK292485.1 | - | 10 - 11 | 1065 | 122 | |
AK307494.1 | - | 11 - 12 | 1241 | 122 | |
AK309772.1 | - | 9 - 10 | 918 | 122 | |
BC034357.1 | - | 10 - 11 | 1046 | 122 | |
BC065481.1 | - | 10 - 11 | 1025 | 122 | |
BC113585.1 | - | 10 - 11 | 1092 | 122 | |
BC113587.1 | - | 10 - 11 | 1092 | 122 | |
BX641140.1 | - | 10 - 11 | 1047 | 122 | |
BX647450.1 | - | 10 - 11 | 1045 | 122 | |
BX647992.1 | - | 7 - 8 | 693 | 122 |