The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177591.1 | NP_001171062.1 | 5 - 6 | 526 | 95 |
NM_002717.3 | NP_002708.1 | 5 - 6 | 794 | 95 | |
XM_005273559.1 | XP_005273616.1 | 5 - 6 | 759 | 95 | |
XM_005273560.1 | XP_005273617.1 | 6 - 7 | 842 | 95 | |
XM_011544565.1 | XP_011542867.1 | 5 - 6 | 1150 | 95 | |
XM_011544566.1 | XP_011542868.1 | 5 - 6 | 781 | 95 | |
XM_011544567.2 | XP_011542869.1 | 6 - 7 | 833 | 95 | |
XM_011544568.1 | XP_011542870.1 | 5 - 6 | 770 | 95 | |
XM_011544569.1 | XP_011542871.1 | 5 - 6 | 502 | 95 | |
GenBank mRNA | AK303981.1 | - | 5 - 6 | 528 | 95 |
AK314823.1 | - | 5 - 6 | 757 | 95 | |
BC041071.1 | - | 5 - 6 | 744 | 95 | |
BX440554.2 | - | 5 - 6 | 768 | 95 | |
M64929.1 | - | 5 - 6 | 570 | 95 |